ContactUsLinkComponent
DL - SULFORAPHANE. Purity: 97% A Phase II enzyme inducer found to inhibit chemically induced mammary tumor formation in rats. 1 * 25 mg
Catalog # ICNA0219378125
Supplier:  MP Biomedicals
DL - SULFORAPHANE. Purity: 97% A Phase II enzyme inducer found to inhibit chemically induced mammary tumor formation in rats. 1 * 25 mg
Catalog # ICNA0219378125
Supplier:  MP Biomedicals
Frequently Bought Together