ContactUsLinkComponent
Purity: 99% Mixed Isomers.Approx. 85% 1,3-isomer and 15% 1,2-isomer. Free of oleic acid. 1 * 50 mg
Catalog # ICNA0219515150
Supplier:  MP Biomedicals
Purity: 99% Mixed Isomers.Approx. 85% 1,3-isomer and 15% 1,2-isomer. Free of oleic acid. 1 * 50 mg
Catalog # ICNA0219515150
Supplier:  MP Biomedicals
Frequently Bought Together