PACLITAXEL. (Taxol®) From the Pacific Yew Tree Taxus brevifolia Antitumor agent which lowers the critical concentration for tubulin polymerization and reversibly binds to tubulin prohibiting depolymerization. Purity: >98% 1 * 100 mg
Catalog # ICNA0219353280
Supplier: MP Biomedicals
Frequently Bought Together