ContactUsLinkComponent
SC 100-S49 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S49, Amb+13 to 100 C, 30-49 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1521491
SC 100-S49 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S49, Amb+13 to 100 C, 30-49 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1521491
Frequently Bought Together