Conductivity probe with integral PT1000 sensor, 2 EL Titanium cell, k= 1 5 µ/cm ... 50 mS/cm, 0 … 80 °C 1 * 1 items
HACHLZW5064970002
: Hach
Conductivity probe with integral PT1000 sensor, 2 EL Titanium cell, k= 1 5 µ/cm ... 50 mS/cm, 0 … 80 °C 1 * 1 items
HACHLZW5064970002
: Hach
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection