ContactUsLinkComponent
Oversleeve#color: blue#size: 44x20#description: latexfree elastic#material: polypropylene#thickness: 30 g/m#packaging: 100 pcs/bag, 1000 pcs/carton 1 * 1.000 items
Catalog # GUNA140133
Supplier:  VWR Collection
Oversleeve#color: blue#size: 44x20#description: latexfree elastic#material: polypropylene#thickness: 30 g/m#packaging: 100 pcs/bag, 1000 pcs/carton 1 * 1.000 items
Catalog # GUNA140133
Supplier:  VWR Collection
Frequently Bought Together