SILICA GEL WITH MOISTURE INDICATOR (ORAN 1 * 170 g
Catalog # 1.43032.0170
Supplier: Merck
Frequently Bought Together
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Kyselina chlorovodíková 2 mol/l (2 N) Reag. Ph. Eur. 1043503
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Dishes, disposable
Supplier: BOCHEM
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Density standard, water
Supplier: Mettler - Toledo
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Measuring cylinders, tall form, class A, borosilicate glass
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Hydroxid sodný 0.1 M NaOH (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Caffeic acid, Supelco®
Supplier: Merck