ContactUsLinkComponent
1000µl Pre-Sterilized Hamilton CO-RE Style Liquid Level Sensing Filter Tip. 96 Tips per Rack, 5 Racks/pack, 8 packs/case 1 * 3.840 items
Catalog # AXYGHTF1000CBKHTS5
Supplier:  Corning
1000µl Pre-Sterilized Hamilton CO-RE Style Liquid Level Sensing Filter Tip. 96 Tips per Rack, 5 Racks/pack, 8 packs/case 1 * 3.840 items
Catalog # AXYGHTF1000CBKHTS5
Supplier:  Corning
Frequently Bought Together