ContactUsLinkComponent
Sieve ring 2 mm trapezoidal perforation, pure titanium with reinforced edges for Variable Speed Rotor Mill PULVERISETTE 14 For heavy duty loads for the heavy-metal-free and iron free grinding of soft materials such as leaves, grains and soft foods. 1 * 1 items
Catalog # FRIT44.3201.32
Supplier:  FRITSCH
Sieve ring 2 mm trapezoidal perforation, pure titanium with reinforced edges for Variable Speed Rotor Mill PULVERISETTE 14 For heavy duty loads for the heavy-metal-free and iron free grinding of soft materials such as leaves, grains and soft foods. 1 * 1 items
Catalog # FRIT44.3201.32
Supplier:  FRITSCH
Frequently Bought Together