ContactUsLinkComponent
Inhibits GSK3β more potently (IC50 = 18 nM) than CDK1/cyclin B or CDK5/p25 (IC50s = 2.0 and 4.2 µM, respectively) 1 * 1 mg
Catalog # CAYM16733-1
Supplier:  Cayman Chemical
Inhibits GSK3β more potently (IC50 = 18 nM) than CDK1/cyclin B or CDK5/p25 (IC50s = 2.0 and 4.2 µM, respectively) 1 * 1 mg
Catalog # CAYM16733-1
Supplier:  Cayman Chemical
Frequently Bought Together