ContactUsLinkComponent
An orally bioavailable, reversible inhibitor of JAK1 and JAK2 (IC50s = 5.9 and 5.7 nM, respectively) that has been shown to inhibit cytokine-induced phosphorylation of STAT3 in healthy human volunteers 1 * 25 mg
Catalog # CAYM16707-25
Supplier:  Cayman Chemical
An orally bioavailable, reversible inhibitor of JAK1 and JAK2 (IC50s = 5.9 and 5.7 nM, respectively) that has been shown to inhibit cytokine-induced phosphorylation of STAT3 in healthy human volunteers 1 * 25 mg
Catalog # CAYM16707-25
Supplier:  Cayman Chemical
Frequently Bought Together