ContactUsLinkComponent
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsi on stabilizers, and surfactants in cosmetics 1 * 1 g
  CAYM10009730-1
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsi on stabilizers, and surfactants in cosmetics 1 * 1 g
  CAYM10009730-1
Frequently Bought Together