ContactUsLinkComponent
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively) 1 * 100 mg
Catalog # CAYM16964-100
Supplier:  Cayman Chemical
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively) 1 * 100 mg
Catalog # CAYM16964-100
Supplier:  Cayman Chemical
Frequently Bought Together