ContactUsLinkComponent
Ex-i power/input isolating amplifier,for the connection ofan ATEX vacuum sensor. Sends fed or active 0/4-20 mA signals from the Ex a rea to a load (active or passive) to the safe area.Electrical 3-way isolation, SIL 2,wide range power supply. 1 * 1 items
Catalog # VACU20635427
Supplier:  VACUUBRAND
Ex-i power/input isolating amplifier,for the connection ofan ATEX vacuum sensor. Sends fed or active 0/4-20 mA signals from the Ex a rea to a load (active or passive) to the safe area.Electrical 3-way isolation, SIL 2,wide range power supply. 1 * 1 items
Catalog # VACU20635427
Supplier:  VACUUBRAND
Frequently Bought Together