ContactUsLinkComponent
ATEX Vacuum gauge head Cerabar T PMP131,Endress+Hauser, with KF DN 16,measuring range: 1000-1 mbaraccuracy: better than 0.5 % FSO,wi th ATEX approval: II 2G Ex ib IIC T6 output signal 4 - 20 mA, for operation with Ex i - power / input isolating ampli 1 * 1 items
Catalog # VACU20635424
Supplier:  VACUUBRAND
ATEX Vacuum gauge head Cerabar T PMP131,Endress+Hauser, with KF DN 16,measuring range: 1000-1 mbaraccuracy: better than 0.5 % FSO,wi th ATEX approval: II 2G Ex ib IIC T6 output signal 4 - 20 mA, for operation with Ex i - power / input isolating ampli 1 * 1 items
Catalog # VACU20635424
Supplier:  VACUUBRAND
Frequently Bought Together