ContactUsLinkComponent
GEL LONG 2% AGAROSE TBE W/ETBR (24+1) 1 * 5 items
Catalog # EMBIGE-4632
Supplier:  EMBITEC
GEL LONG 2% AGAROSE TBE W/ETBR (24+1) 1 * 5 items
Catalog # EMBIGE-4632
Supplier:  EMBITEC
Frequently Bought Together