ContactUsLinkComponent
BSA Protein Digest Standard, LC-MS Grade, is a validated protein digest optimized for use as a quality control sample for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples. 1 * 1 Nmo
Catalog # PIER88341
undefined
BSA Protein Digest Standard, LC-MS Grade, is a validated protein digest optimized for use as a quality control sample for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples. 1 * 1 Nmo
Catalog # PIER88341
Frequently Bought Together