ContactUsLinkComponent
SONOREX SUPER RK 255 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 300X150X150 MM 5,5 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEA TING 30-80°C (86-176°F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND316-CH
SONOREX SUPER RK 255 H, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 300X150X150 MM 5,5 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEA TING 30-80°C (86-176°F), DRAIN BALL COCK 1/4 , ULTRASONIC PEAK OUTPUT 640 W, HF-OUTPUT: 160 WEFF. 1 * 1 items
Catalog # BAND316-CH
Frequently Bought Together