SONOREX SUPER RK 52, ULTRASONIC BATH, 35 KHZ, 230 V~ 50/60 HZ, ID 150X140X100 MM 1,8 L, S/S TIMER 1-15 MIN AND CONTINUOUS, ULTRAS ONIC PEAK OUTPUT 240 W, HF-OUTPUT: 60 WEFF. 1 * 1 items
Catalog # BAND311-CH
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together