ContactUsLinkComponent
SONOREX TECHNIK RM 180 UH, 40 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 1000X500X400 MM 200 L, WELDED TANK 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 2X1000 WEFF., WITH OVERFL 1 * 1 items
Catalog # BAND8250-GB
SONOREX TECHNIK RM 180 UH, 40 KHZ, ULTRASONIC BATH, 400 V 3N 50/60 HZ, ID 1000X500X400 MM 200 L, WELDED TANK 2 MM S/S AISI 316 TI TIMER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 4000 W, HF-OUTPUT: 2X1000 WEFF., WITH OVERFL 1 * 1 items
Catalog # BAND8250-GB
Frequently Bought Together