ContactUsLinkComponent
SONOREX TECHNIK RM 16 UH, ULTRASONIC BATH, 40 KHZ, 230 V 50/60 HZ, ID 325X275X200 MM 17,8 L, WELDED TANK 2 MM S/S AISI 316 TI TI MER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 1X300 WEFF., WITH OVERFLOW GU 1 * 1 items
Catalog # BAND8200-CH
SONOREX TECHNIK RM 16 UH, ULTRASONIC BATH, 40 KHZ, 230 V 50/60 HZ, ID 325X275X200 MM 17,8 L, WELDED TANK 2 MM S/S AISI 316 TI TI MER 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 1200 W, HF-OUTPUT: 1X300 WEFF., WITH OVERFLOW GU 1 * 1 items
Catalog # BAND8200-CH
Frequently Bought Together