ContactUsLinkComponent
SONOREX SUPER RK 1050, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 600X500X200 MM 58 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 2400 W, HF-OUTPUT: 600 WEFF. 1 * 1 items
Catalog # BAND323-GB
SONOREX SUPER RK 1050, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 600X500X200 MM 58 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAIN BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 2400 W, HF-OUTPUT: 600 WEFF. 1 * 1 items
Catalog # BAND323-GB
Frequently Bought Together