ContactUsLinkComponent
Microplate, 96 Well Stripwell, 8 Well Strips, Polystyrene, White, Flat Bottom, High Binding, no Lid, Nonsterile, Bulk 1 * 100 items
Catalog # CORN93923
Supplier:  Corning
Microplate, 96 Well Stripwell, 8 Well Strips, Polystyrene, White, Flat Bottom, High Binding, no Lid, Nonsterile, Bulk 1 * 100 items
Catalog # CORN93923
Supplier:  Corning
Frequently Bought Together