ContactUsLinkComponent
Microplate, 96 Well Stripwell, 8 Well Strips, Polystyrene, Clear, Flat Bottom, High Binding, no Lid, Red Hot Stamp Foil, Nonsterile , Bulk 1 * 100 items
Catalog # CORN9459
Supplier:  Corning
Microplate, 96 Well Stripwell, 8 Well Strips, Polystyrene, Clear, Flat Bottom, High Binding, no Lid, Red Hot Stamp Foil, Nonsterile , Bulk 1 * 100 items
Catalog # CORN9459
Supplier:  Corning
Frequently Bought Together