CLOZAPINE PEAK IDENTIFICATION 1 * 0,41 mg
Catalog # EDQMY0000758
Supplier: EDQM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Viscosity standards
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals