ASPARTAME 1 * 40 mg
Catalog # EDQMA1320000
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Bottles, Wide Neck with Screw Cap
Supplier: RIXIUS