ContactUsLinkComponent
Llama IgG Purified contains Protein G purified llama IgG from normal serum, e.g. serum of non immunized animals and is excellent fo r use as blocking reagent in immunoassays.,Llama IgG Purified contains Protein G purified llama IgG from normal serum 1 * 10 mg
Catalog # AGRIAS152844
Supplier:  Agrisera
Llama IgG Purified contains Protein G purified llama IgG from normal serum, e.g. serum of non immunized animals and is excellent fo r use as blocking reagent in immunoassays.,Llama IgG Purified contains Protein G purified llama IgG from normal serum 1 * 10 mg
Catalog # AGRIAS152844
Supplier:  Agrisera