ContactUsLinkComponent
With a total measuring range of 0.01 – 2100 µm in a single instrument, the ANALYSETTE 22 NanoTec is the ideal, universally applicable Laser Particle Sizer for the effective and reliable determination of particle size distributions down to the nano r 1 * 1 items
Catalog # FRIT22.8000.00
Supplier:  FRITSCH
With a total measuring range of 0.01 – 2100 µm in a single instrument, the ANALYSETTE 22 NanoTec is the ideal, universally applicable Laser Particle Sizer for the effective and reliable determination of particle size distributions down to the nano r 1 * 1 items
Catalog # FRIT22.8000.00
Supplier:  FRITSCH
Frequently Bought Together