ContactUsLinkComponent
The bottom sieves are made of stainless steel trapezoidal resp. round perforation sheets metal. Larger openings mean faster throughput, less time in the mill and less wear. Bottom sieves with trapezoidal perforation effect improved size reduction th 1 * 1 items
Catalog # FRIT16.6724.10
Supplier:  FRITSCH
The bottom sieves are made of stainless steel trapezoidal resp. round perforation sheets metal. Larger openings mean faster throughput, less time in the mill and less wear. Bottom sieves with trapezoidal perforation effect improved size reduction th 1 * 1 items
Catalog # FRIT16.6724.10
Supplier:  FRITSCH
Frequently Bought Together