ContactUsLinkComponent
Cross Beater Mill PULVERISETTE 16 cast iron grinding insert Suitable for fast and gentle pre-crushing and fine comminution of medium-hard, brittle materials up to a Mohs hardness of 6. Max. feed size: (depends on material) 20 mm Max. throughput quant 1 * 1 items
Catalog # FRIT16.6020.00
Supplier:  FRITSCH
Cross Beater Mill PULVERISETTE 16 cast iron grinding insert Suitable for fast and gentle pre-crushing and fine comminution of medium-hard, brittle materials up to a Mohs hardness of 6. Max. feed size: (depends on material) 20 mm Max. throughput quant 1 * 1 items
Catalog # FRIT16.6020.00
Supplier:  FRITSCH
Frequently Bought Together