Density hydrometer No. 11, 1.300-1.360 without thermometer, 160 mm long 1 * 1 items
AMARH800024/KB
: Amarell
Density hydrometer No. 11, 1.300-1.360 without thermometer, 160 mm long 1 * 1 items
AMARH800024/KB
: Amarell
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK