CALCIUM CONC. 1,000±0,002 IN 2% HNO3 1 * 100 mL
Catalog # ANLYCZ90091N1
Supplier: ANALYTIKA
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions
Supplier: REAGECON
IRON CONC. 1,000±0,002 IN 2% HNO3 1 * 100 mL
Supplier: ANALYTIKA
pH Buffer solutions
Supplier: Hach
SULPHATES CONC. 1,000±0,002 IN H2O 1 * 500 mL
Supplier: ANALYTIKA
pH Standard Solutions, IUPAC, Certified
Supplier: Hach