AMMONIUM CONC. 1,000±0,002 IN H2O 1 * 100 mL
Catalog # ANLYCZ90801H1
Supplier: ANALYTIKA
Frequently Bought Together
AVENANTHRAMIDE D, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
NITRATES CONC. 1,000±0,002 IN H2O 1 * 500 mL
Supplier: ANALYTIKA
BISOPROLOL FOR SYSTEM SUITABILITY 1 * 1 mg
Supplier: EDQM
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
CALCIUM CONC. 1,000±0,002 IN 2% HNO3 1 * 100 mL
Supplier: ANALYTIKA
Organic reference standards, Chloroparaffin C10-C13, Dr. Ehrenstorfer
Supplier: EHRENSTORFER
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Paracetamol
Supplier: EDQM
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
Supelco® Organic Reference Standard, Diallyl Phthalate
Supplier: Merck
MENADIONE 1 * 100 mg
Supplier: EDQM
Supelco® Organic Reference Standard, α-Arbutin
Supplier: Merck
Pesticide reference standard, Endrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
3-NITROSALICYLIC ACID 1 * 100 mg
Supplier: Merck