SULPHATES CONC. 1,000±0,002 IN H2O 1 * 500 mL
Catalog # ANLYCZ90781H5
Supplier: ANALYTIKA
Frequently Bought Together
Baňka titrační,250ml,SIMAX 1 * 1 KS
Supplier: Kavalier
Malachite green solution Reag. Ph. Eur. 1050501
Supplier: VWR Chemicals
PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
Supelco® Organic Reference Standard, Nonylphenol Solution
Supplier: Merck
Sulfite lr tablets in pack of 100 1 * 100 Tablet
Supplier: Lovibond
Thiochrome, analytical standard, Supelco®
Supplier: Merck
Alachlor ethane sulfonic acid (ESA) sodium salt certified 1 * 10 mg
Supplier: EHRENSTORFER
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck
Sulfalene 1 * 100 mg
Supplier: EHRENSTORFER
AMMONIUM CONC. 1,000±0,002 IN H2O 1 * 100 mL
Supplier: ANALYTIKA
ALUMINIUM CONC. 1,000±0,002 IN 5% HNO3 1 * 100 mL
Supplier: ANALYTIKA
Buffer Solutions
Supplier: Thermo Fisher Scientific
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
POLYETHYLENE OXIDE CALIBRATION KIT 1 * 1 items
Supplier: VARIAN
Clothianidin certified 1 * 0,1 g
Supplier: EHRENSTORFER