PHOSPHATES CONC. 1,000±0,002 IN H2O 1 * 100 mL
Catalog # ANLYCZ90771H1
Supplier: ANALYTIKA
Frequently Bought Together
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
VWR®, AVS TITRINORM pH pufrovací roztoky v sáčcích, 20 °C
Supplier: VWR Chemicals
Aluminium foil
Supplier: Alujet-Universal
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Roztoky standardní pro měření vodivosti
Supplier: VWR Chemicals
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals