MAGNESIUM CONC. 10,00±0,02 IN 2% HNO3 1 * 100 mL
Catalog # ANLYCZ903210N1
Supplier: ANALYTIKA
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Glass Fibre Filters
Supplier: VWR Collection
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
ALUMINIUM CONC. 1,000±0,002 IN 5% HNO3 1 * 100 mL
Supplier: ANALYTIKA