IRON CONC. 1,000±0,002 IN 2% HNO3 1 * 100 mL
Catalog # ANLYCZ90191N1
Supplier: ANALYTIKA
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Accessories for pH/°C meter, handheld
Supplier: Testo
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
NITRATES CONC. 1,000±0,002 IN H2O 1 * 500 mL
Supplier: ANALYTIKA
MAGNESIUM CONC. 10,00±0,02 IN 2% HNO3 1 * 100 mL
Supplier: ANALYTIKA
pH Standard Solutions, IUPAC, Certified
Supplier: Hach