ContactUsLinkComponent
Interleukin-22 (IL-22) is a member of a group of cytokines called the IL-10 family which include IL-10,IL-19, IL-20, IL-24, and IL-2 6. IL-22 shares use of the IL-10R2 in cell signaling with other members of this family IL-22 signals through a recept 1 * 10 µG
Catalog # GENSZ03315-10
Supplier:  Genscript
Interleukin-22 (IL-22) is a member of a group of cytokines called the IL-10 family which include IL-10,IL-19, IL-20, IL-24, and IL-2 6. IL-22 shares use of the IL-10R2 in cell signaling with other members of this family IL-22 signals through a recept 1 * 10 µG
Catalog # GENSZ03315-10
Supplier:  Genscript
Frequently Bought Together