Disposable syringes
Luer lock syringe, 20 ml
Catalog # NSCAS7515-20
Supplier: THERMO NATIONAL SCIENTIFIC
Jump to:
Product Details & Documents
These syringes are an ideal choice for all syringe filter applications. They feature a two-part construction using PP barrels and PE plungers. This eliminates the need for rubber or synthetic plunger gaskets and requires no silicone or oil lubricant in the barrel.
- Non sterile
- Bulk packed
Specifications
- Volume:20 ml
- Description:Luer lock syringe
- Seq:04