MOLYBDATE ION CHROMATOGRAPHY STANDARD IN H2O MOLYBDATE - MOO4(-2) @1000µG/ML 1 * 100 mL
Catalog # PROOVHG-IMOLB-100
Supplier: LGC Standards PROMOCHEM
MOLYBDATE ION CHROMATOGRAPHY STANDARD IN H2O MOLYBDATE - MOO4(-2) @1000µG/ML 1 * 100 mL
Catalog # PROOVHG-IMOLB-100
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR® ARISTAR®, Aluminium Standard Solutions for ICP
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Supplier: VWR Chemicals
Polystyrene, analytical standards for GPC
Supplier: Merck
BUFFER SOLUTION PH 4.01 25C 1 * 500 mL
Supplier: REAGECON
Nitralin certified 1 * 0,1 g
Supplier: EHRENSTORFER
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, Protein Standard
Supplier: Merck
Triethyl Citrate, Reference Standard
Supplier: USP
Tioxazafen 1 * 10 mg
Supplier: EHRENSTORFER