US EPA METHOD 200.7 ICP TUNING SOLUTION @ 10µG/ML IN 5% HNO3 1 * 500 mL
PROOVHG-TNG-500
US EPA METHOD 200.7 ICP TUNING SOLUTION @ 10µG/ML IN 5% HNO3 1 * 500 mL
PROOVHG-TNG-500
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection