ContactUsLinkComponent
Measuring Jug, 2000 mL PP, Materials supplied by: EP. Process: Particulate Cleaning. Packaging: Double autoclave bag each cylinder. Place autoclave bag in a single cleanroom bag. 4 per case in original box. Note: Item is packaged for autoclave but ha 1 * 4 items
Catalog # NALG7LVNL2P-EP
Measuring Jug, 2000 mL PP, Materials supplied by: EP. Process: Particulate Cleaning. Packaging: Double autoclave bag each cylinder. Place autoclave bag in a single cleanroom bag. 4 per case in original box. Note: Item is packaged for autoclave but ha 1 * 4 items
Catalog # NALG7LVNL2P-EP
Frequently Bought Together