ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, V, P+G, WB, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Vertical (V) P+G coating, int.: Wou lff b., w/o int. 1 * 1 items
Catalog # BUCH11R300252V111
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, V, P+G, WB, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Vertical (V) P+G coating, int.: Wou lff b., w/o int. 1 * 1 items
Catalog # BUCH11R300252V111
Supplier:  BUCHI
Frequently Bought Together