ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, no glass, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: w /o int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300152N011
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, no glass, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: w /o int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300152N011
Supplier:  BUCHI
Frequently Bought Together