ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, C, P+G, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Col d trap, P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300151C111
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, C, P+G, WB, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: Col d trap, P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300151C111
Supplier:  BUCHI
Frequently Bought Together