ContactUsLinkComponent
Rotavapor R-300 w coated diag cond ass and manual jack. Heating bath B-305 with a digital display and temprange to 220°C. Max flask size 5000mL. Evap flask 1L. Rec flask 1L. Standard joint. SJ 29/32. 1 * 1 items
Catalog # BUCH11R300151A101
Supplier:  BUCHI
Rotavapor R-300 w coated diag cond ass and manual jack. Heating bath B-305 with a digital display and temprange to 220°C. Max flask size 5000mL. Evap flask 1L. Rec flask 1L. Standard joint. SJ 29/32. 1 * 1 items
Catalog # BUCH11R300151A101
Supplier:  BUCHI
Frequently Bought Together