ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, S, P+G, WB, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: Vert ical reflux (S) P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300112S111
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, S, P+G, WB, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: Vert ical reflux (S) P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300112S111
Supplier:  BUCHI
Frequently Bought Together