ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, A, P+G, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Diagonal (A) P+G coating, 1 * 1 items
Katalogové číslo BUCH11R300111A101
Dodavatel:  BUCHI
Rotavapor R-300 B-301, SJ29/32, A, P+G, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Diagonal (A) P+G coating, 1 * 1 items
Katalogové číslo BUCH11R300111A101
Dodavatel:  BUCHI
Frequently Bought Together