ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, S, P+G, WB, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical reflux (S) P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300101S111
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, S, P+G, WB, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: Vertical reflux (S) P+G coating, int.: Woulff b., w/o int. 1 * 1 items
Catalog # BUCH11R300101S111
Supplier:  BUCHI
Frequently Bought Together