ContactUsLinkComponent
Rotavapor R-300 no bath, SJ24/40, A, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 24/40 Glass set: Diagonal (A) int. : I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300202A051
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ24/40, A, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 24/40 Glass set: Diagonal (A) int. : I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300202A051
Supplier:  BUCHI
Frequently Bought Together